Learn More
Medchemexpress LLC HY-19363 10mg Medchemexpress, GW4869 CAS:6823-69-4 Purity:>98%
SDP
Catalog No. 502029279
Encompass_Preferred
Medchemexpress, HY-19363 10mg GW4869 CAS:6823-69-4 GW4869 is a noncompetitive neutral sphingomyelinase (N-SMase) inhibitor with an IC50 of 1 μM. GW4869 is an inhibitor of exosome biogenesis/release[2][3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9279 Supplier Medchemexpress LLC Supplier No. HY1936310MG
Medchemexpress, HY-19363 10mg GW4869 CAS:6823-69-4 GW4869 is a noncompetitive neutral sphingomyelinase (N-SMase) inhibitor with an IC50 of 1 μM. GW4869 is an inhibitor of exosome biogenesis/release[2][3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.