Learn More
Selleck Chemical LLC GW4869 5mg 6823-69-4 GW69A, GW554869A
SDP
Supplier:  Selleck Chemical LLC S76095MG
Encompass_Preferred
GW4869 (GW69A, GW554869A) is a neutral, noncompetitive inhibitor of sphingomyelinase (SMase) with an IC50 of 1 ?M. It is selective for N-SMase, and does not inhibit acid SMase at up to at least 150 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5019