Learn More
Medchemexpress LLC Gw4869 | 6823-69-4 | 98.9% | 577.50 | 250 MG
SDP
Catalog No. 5000384198
Encompass_Preferred
GW4869 is a noncompetitive neutral sphingomyelinase (N-SMase) inhibitor with an IC50 of 1 μM. It also acts as an inhibitor of exosome biogenesis and release. This product is intended for research use only.
  • Noncompetitive neutral sphingomyelinase (N-SMase) inhibitor
  • Inhibits exosome biogenesis and release
  • Partially inhibits TNF-induced sphingomyelin (SM) hydrolysis
  • Protects from cell death in a dose-dependent manner
  • Reduces pro-inflammatory cytokine production in macrophages
  • Blocks ceramide-mediated inward budding of multivesicular bodies (MVBs)
  • In vivo, inhibits exosome release and mitigates LPS-caused myocardial dysfunction
Catalog No. 50-003-84198 Supplier Medchemexpress LLC Supplier No. 250MGHY19363
GW4869 is a noncompetitive neutral sphingomyelinase (N-SMase) inhibitor with an IC50 of 1 μM. It also acts as an inhibitor of exosome biogenesis and release. This product is intended for research use only.
  • Noncompetitive neutral sphingomyelinase (N-SMase) inhibitor
  • Inhibits exosome biogenesis and release
  • Partially inhibits TNF-induced sphingomyelin (SM) hydrolysis
  • Protects from cell death in a dose-dependent manner
  • Reduces pro-inflammatory cytokine production in macrophages
  • Blocks ceramide-mediated inward budding of multivesicular bodies (MVBs)
  • In vivo, inhibits exosome release and mitigates LPS-caused myocardial dysfunction