Learn More
Enzo Life Sciences GW501516 (5mg). CAS: 317318-70-0
SDP
Catalog No. 502009109
Encompass_Preferred
GW501516 is a specific agonist for PPARδ (EC50=1.1nM) with a 1'000-fold selectivity over the other human subtypes. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (50mg/ml), dimethyl formamide (25mg/ml), 100% ethanol (12mg/ml) or methanol (50mg/ml); slightly soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9109 Supplier Enzo Life Sciences Supplier No. ALX420032M005
GW501516 is a specific agonist for PPARδ (EC50=1.1nM) with a 1'000-fold selectivity over the other human subtypes. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (50mg/ml), dimethyl formamide (25mg/ml), 100% ethanol (12mg/ml) or methanol (50mg/ml); slightly soluble in water. Long Term Storage: +4°C.