Learn More
Selleck Chemical LLC GW5074 5mg 220904-83-6
SDP
Supplier:  Selleck Chemical LLC S28725MG
Encompass_Preferred
GW5074 is a potent and selective c-Raf inhibitor with IC50 of 9 nM, no effect on the activities of JNK1/2/3, MEK1, MKK6/7, CDK1/2, c-Src, p38 MAP, VEGFR2 or c-Fms is noted. GW5074 inhibits LK-induced apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3115