Learn More
Apexbio Technology LLC GW5074, 5mg. Cas: 220904-83-6 MFCD: MFCD03453076. C-Raf inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC B14055
Encompass_Preferred
GW5074 is a synthetic inhibitor of c-Raf with IC50 value of 9nM [1].GW5074 is found to be a potent inhibitor of c-Raf and has no effect on cdk1, cdk2, c-src, p38 MAP kinase, VEGFR2 and c-fms. GW5074 also shows no direct effect on JNK proteins and p38 MAP kinase. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5229