Learn More
Abcam GW542573 x , KCa2 channel activator, 10MG
Catalog No. 502707544
Encompass_Preferred
MW 364.4 Da, Purity >99%. Novel activator of KCa2 ('SK') channels. Selective SK1 channel activator (EC50 = 8.2 μM). First true activator of KCa2.x and KCa3.1 channels; displays 100-fold greater selectivity for KCa2.x over KCa3.1.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7544 Supplier Abcam Supplier No. AB12050210MG
MW 364.4 Da, Purity >99%. Novel activator of KCa2 ('SK') channels. Selective SK1 channel activator (EC50 = 8.2 μM). First true activator of KCa2.x and KCa3.1 channels; displays 100-fold greater selectivity for KCa2.x over KCa3.1.
The product is subject to the following: Abcam Restricted Use Statement