Learn More
Medchemexpress LLC HY-138559 5mg , GW604714X CAS:853953-65-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1385595MG
Encompass_Preferred
HY-138559 5mg GW604714X CAS: 853953-65-8 GW604714X is a potent inhibitor of mitochondrial respiration supported by pyruvate but not other substrates. GW604714X is a highly specific mitochondrial pyruvate carrier (MPC) inhibitor with a Ki <0.1 nM. GW604714X also inhibits L-lactate transport by the plasma membrane monocarboxylate transporter (MCT1), but at concentrations more than 4 orders of magnitude greater than the MPC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1425