Learn More
Medchemexpress LLC GW604714X | 853953-65-8 | 98.5% | 471.46 | 100 MG
SDP
Catalog No. 5000352365
Encompass_Preferred
GW604714X is a potent inhibitor of mitochondrial respiration supported by pyruvate but not other substrates. It is a highly specific mitochondrial pyruvate carrier (MPC) inhibitor with a Ki value of less than 0.1 nM. Additionally, GW604714X can inhibit L-lactate transport by the plasma membrane monocarboxylate transporter (MCT1), although this occurs at concentrations more than 4 orders of magnitude higher than those required for MPC inhibition.
  • Potent inhibitor of mitochondrial respiration
  • Highly specific mitochondrial pyruvate carrier (MPC) inhibitor with a Ki <0.1 nM
  • Inhibits L-lactate transport by the plasma membrane monocarboxylate transporter (MCT1) at higher concentrations
  • Inhibits mitochondrial pyruvate carrier (MPC) with a Ki value <0.1 μM in direct measurement of pyruvate transport
Catalog No. 50-003-52365 Supplier Medchemexpress LLC Supplier No. HY138559100MG
GW604714X is a potent inhibitor of mitochondrial respiration supported by pyruvate but not other substrates. It is a highly specific mitochondrial pyruvate carrier (MPC) inhibitor with a Ki value of less than 0.1 nM. Additionally, GW604714X can inhibit L-lactate transport by the plasma membrane monocarboxylate transporter (MCT1), although this occurs at concentrations more than 4 orders of magnitude higher than those required for MPC inhibition.
  • Potent inhibitor of mitochondrial respiration
  • Highly specific mitochondrial pyruvate carrier (MPC) inhibitor with a Ki <0.1 nM
  • Inhibits L-lactate transport by the plasma membrane monocarboxylate transporter (MCT1) at higher concentrations
  • Inhibits mitochondrial pyruvate carrier (MPC) with a Ki value <0.1 μM in direct measurement of pyruvate transport