Learn More
Selleck Chemical LLC GW6471 10mg
SDP
Supplier:  Selleck Chemical LLC S279810MG
Encompass_Preferred
GW 6471 is a potent antagonist of PPAR? with IC50 of 0.24 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3104