Learn More
Medchemexpress LLC GW6471 | 880635-03-0 | C35H36F3N3O4 | 100 MG
SDP
Catalog No. 5000376042
Encompass_Preferred
GW6471 is a potent PPARα antagonist that inhibits GW409544-induced activation of PPARα with an IC50 of 0.24 μM. It significantly inhibits cell viability in Caki-1 and 786-O renal cell carcinoma cell lines in vitro. In vivo studies demonstrate its antitumor activity, showing decreased tumor growth and c-Myc levels in xenograft mouse models without observed toxicity.
  • Potent PPARα antagonist
  • Inhibits cell viability in renal cell carcinoma cell lines
  • Demonstrates antitumor activity in vivo
  • No toxicity observed at effective doses in animal studies
  • Cited in numerous publications, including Nat Metab, Cell Metab, and Immunity
This item is not returnable. View return policy
Catalog No. 50-003-76042 Supplier Medchemexpress LLC Supplier No. HY15372100MG
GW6471 is a potent PPARα antagonist that inhibits GW409544-induced activation of PPARα with an IC50 of 0.24 μM. It significantly inhibits cell viability in Caki-1 and 786-O renal cell carcinoma cell lines in vitro. In vivo studies demonstrate its antitumor activity, showing decreased tumor growth and c-Myc levels in xenograft mouse models without observed toxicity.
  • Potent PPARα antagonist
  • Inhibits cell viability in renal cell carcinoma cell lines
  • Demonstrates antitumor activity in vivo
  • No toxicity observed at effective doses in animal studies
  • Cited in numerous publications, including Nat Metab, Cell Metab, and Immunity