Learn More
Medchemexpress LLC HY-117153 5mg Medchemexpress, GW7604 CAS:195611-82-6 Purity:>98%
SDP
Catalog No. 502080899
Encompass_Preferred
Medchemexpress, HY-117153 5mg GW7604 CAS:195611-82-6 GW7604 is an antiestrogen. GW7604 is the metabolite of GW5638, which is a high affinity estrogen receptor (ER) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0899 Supplier Medchemexpress LLC Supplier No. HY1171535MG
Medchemexpress, HY-117153 5mg GW7604 CAS:195611-82-6 GW7604 is an antiestrogen. GW7604 is the metabolite of GW5638, which is a high affinity estrogen receptor (ER) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.