Learn More
Abcam GW7647, PPARalpha agonist, 50MG
Catalog No. 502706879
Encompass_Preferred
MW 502.8 Da, Purity >99%. Highly potent, selective PPARα agonist (EC₅₀ values are 6, 1100 and 6200 nM for PPARα, PPARγ and PPARδ respectively). Pharmacologically active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6879 Supplier Abcam Supplier No. AB14112450MG
MW 502.8 Da, Purity >99%. Highly potent, selective PPARα agonist (EC₅₀ values are 6, 1100 and 6200 nM for PPARα, PPARγ and PPARδ respectively). Pharmacologically active in vivo.
The product is subject to the following: Abcam Restricted Use Statement