Learn More
Medchemexpress LLC HY-13861 100mg Medchemexpress, GW7647 CAS:265129-71-3 Purity:98%
SDP
Catalog No. 501638054
Encompass_Preferred
Medchemexpress,HY-13861 100mg GW7647 CAS:265129-71-3 Formula:C29H46N2O3S GW7647 is a potent PPARα agonist, with EC50s of 6 nM, 1.1 μM, and 6.2 μM for human PPARα, PPARγ and PPARδ, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8054 Supplier Medchemexpress LLC Supplier No. HY13861100MG
Medchemexpress,HY-13861 100mg GW7647 CAS:265129-71-3 Formula:C29H46N2O3S GW7647 is a potent PPARα agonist, with EC50s of 6 nM, 1.1 μM, and 6.2 μM for human PPARα, PPARγ and PPARδ, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.