Learn More
TARGETMOL CHEMICALS INC GW788388 25MG
SDP
Catalog No. 502581090
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GW788388 is a potent and selective inhibitor of ALK5 also inhibits TGF-(beta) type II receptor and activin type II receptor activities. purity: 98%
Catalog No. 50-258-1090 Supplier TARGETMOL CHEMICALS INC Supplier No. T180025MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GW788388 is a potent and selective inhibitor of ALK5 also inhibits TGF-(beta) type II receptor and activin type II receptor activities. purity: 98%