Learn More
Selleck Chemical LLC GW788388 5mg
SDP
Supplier:  Selleck Chemical LLC S27505MG
Encompass_Preferred
GW788388 is a potent and selective inhibitor of ALK5 with IC50 of 18 nM in a cell-free assay, also inhibits TGF-? type II receptor and activin type II receptor activities, but does not inhibit BMP type II receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3102