Learn More
Apexbio Technology LLC GW788388, 452342-67-5, MFCD16038936, 5 mg
SDP
Supplier:  Apexbio Technology LLC A83015
Encompass_Preferred
MF: C25H23N5O2, MW: 425.49, Solubility: 21.25mg/mL in DMSO,insoluble in EtOH, insoluble in H2O. GW788388 is a selective inhibitor of ALK5 with IC50 value of 18 nM.
Catalog No. 50-101-2853