Learn More
Medchemexpress LLC HY-112292 10mg Medchemexpress, GW806742X CAS:579515-63-2 Purity:>98%
SDP
Catalog No. 501969074
Encompass_Preferred
Medchemexpress, HY-112292 10mg GW806742X CAS:579515-63-2 GW806742X is a Mixed Lineage Kinase Domain-Like protein (MLKL) inhibitor which binds the MLKL pseudokinase domain with a Kd value of 9.3 μM and has anti-necroptosis activity. GW806742X has activity against VEGFR2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9074 Supplier Medchemexpress LLC Supplier No. HY11229210MG
Medchemexpress, HY-112292 10mg GW806742X CAS:579515-63-2 GW806742X is a Mixed Lineage Kinase Domain-Like protein (MLKL) inhibitor which binds the MLKL pseudokinase domain with a Kd value of 9.3 μM and has anti-necroptosis activity. GW806742X has activity against VEGFR2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.