Learn More
TARGETMOL CHEMICALS INC GW806742X 25MG
SDP
Catalog No. 502577453
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GW806742X is an inhibitor of Mixed Lineage Kinase Domain-Like protein (MLKL) which binds the MLKL pseudokinase domain (Kd 9.3 uM) with anti-necroptosis activity. GW806742X has activity against VEGFR2. purity: 99%
Catalog No. 50-257-7453 Supplier TARGETMOL CHEMICALS INC Supplier No. T1152025MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GW806742X is an inhibitor of Mixed Lineage Kinase Domain-Like protein (MLKL) which binds the MLKL pseudokinase domain (Kd 9.3 uM) with anti-necroptosis activity. GW806742X has activity against VEGFR2. purity: 99%