Learn More
Medchemexpress LLC GW806742X hydrochloride | 98.3% | 610.01 | 100 MG
SDP
Catalog No. 5000438398
Encompass_Preferred
GW806742X hydrochloride is an ATP mimetic and a potent inhibitor of MLKL (Mixed Lineage Kinase Domain-Like protein), binding to the MLKL pseudokinase domain with a Kd of 9.3 μM. It also exhibits activity against VEGFR2 with an IC50 of 2 nM. This compound has been shown to retard MLKL membrane translocation and inhibit necroptosis.
  • Retards MLKL membrane translocation
  • Inhibits necroptosis
  • Inhibits necroptotic death of wild-type mouse dermal fibroblasts (MDFs) stimulated with TSQ in a dose-dependent manner
  • Inhibits VEGF-induced proliferation of HUVECs with an IC50 of 5 nM
  • Binds MLKL pseudokinase domain with a Kd of 9.3 μM
  • Activity against VEGFR2 with an IC50 of 2 nM
Catalog No. 50-004-38398 Supplier Medchemexpress LLC Supplier No. HY112292A100MG
GW806742X hydrochloride is an ATP mimetic and a potent inhibitor of MLKL (Mixed Lineage Kinase Domain-Like protein), binding to the MLKL pseudokinase domain with a Kd of 9.3 μM. It also exhibits activity against VEGFR2 with an IC50 of 2 nM. This compound has been shown to retard MLKL membrane translocation and inhibit necroptosis.
  • Retards MLKL membrane translocation
  • Inhibits necroptosis
  • Inhibits necroptotic death of wild-type mouse dermal fibroblasts (MDFs) stimulated with TSQ in a dose-dependent manner
  • Inhibits VEGF-induced proliferation of HUVECs with an IC50 of 5 nM
  • Binds MLKL pseudokinase domain with a Kd of 9.3 μM
  • Activity against VEGFR2 with an IC50 of 2 nM