Learn More
Medchemexpress LLC GW806742X hydrochloride | 98.3% | 610.01 | 25 MG
SDP
Catalog No. 5000439627
Encompass_Preferred
GW806742X hydrochloride is an ATP mimetic and a potent MLKL inhibitor that binds the MLKL pseudokinase domain with a Kd of 9.3 μM. It also exhibits activity against VEGFR2 (IC50=2 nM). This compound retards MLKL membrane translocation and inhibits necroptosis. It is for research use only.
  • Potent MLKL inhibitor (Kd of 9.3 μM)
  • Active against VEGFR2 (IC50=2 nM)
  • Retards MLKL membrane translocation
  • Inhibits necroptosis
Catalog No. 50-004-39627 Supplier Medchemexpress LLC Supplier No. 25MGHY112292A
GW806742X hydrochloride is an ATP mimetic and a potent MLKL inhibitor that binds the MLKL pseudokinase domain with a Kd of 9.3 μM. It also exhibits activity against VEGFR2 (IC50=2 nM). This compound retards MLKL membrane translocation and inhibits necroptosis. It is for research use only.
  • Potent MLKL inhibitor (Kd of 9.3 μM)
  • Active against VEGFR2 (IC50=2 nM)
  • Retards MLKL membrane translocation
  • Inhibits necroptosis