Learn More
Medchemexpress LLC GW806742X hydrochloride | 98.3% | 610.01 | 5 MG
SDP
Catalog No. 5000365967
Encompass_Preferred
GW806742X hydrochloride is an ATP mimetic and a potent MLKL (Mixed Lineage Kinase Domain-Like protein) inhibitor. It binds the MLKL pseudokinase domain with a Kd of 9.3 μM and exhibits activity against VEGFR2 with an IC50 of 2 nM. This compound retards MLKL membrane translocation and inhibits necroptosis. This product is for research use only.
  • Potent MLKL inhibitor
  • Activity against VEGFR2 (IC50=2 nM)
  • Retards MLKL membrane translocation
  • Inhibits necroptosis
Catalog No. 50-003-65967 Supplier Medchemexpress LLC Supplier No. 5MGHY112292A
GW806742X hydrochloride is an ATP mimetic and a potent MLKL (Mixed Lineage Kinase Domain-Like protein) inhibitor. It binds the MLKL pseudokinase domain with a Kd of 9.3 μM and exhibits activity against VEGFR2 with an IC50 of 2 nM. This compound retards MLKL membrane translocation and inhibits necroptosis. This product is for research use only.
  • Potent MLKL inhibitor
  • Activity against VEGFR2 (IC50=2 nM)
  • Retards MLKL membrane translocation
  • Inhibits necroptosis