Learn More
Medchemexpress LLC HY-14167 5mg Medchemexpress, GW842166X CAS:666260-75-9 Purity:>98%
SDP
Catalog No. 502081568
Encompass_Preferred
Medchemexpress, HY-14167 5mg GW842166X CAS:666260-75-9 GW842166X is a potent and selective cannabinoid receptor 2 (CB2) agonist with IC50 values of 63 and 91 nM for human and rat CB2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1568 Supplier Medchemexpress LLC Supplier No. HY141675MG
Medchemexpress, HY-14167 5mg GW842166X CAS:666260-75-9 GW842166X is a potent and selective cannabinoid receptor 2 (CB2) agonist with IC50 values of 63 and 91 nM for human and rat CB2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.