Learn More
Selleck Chemical LLC GW9662 10mg 22978-25-2
SDP
Supplier:  Selleck Chemical LLC S291510MG
Encompass_Preferred
GW9662 is a selective PPAR antagonist for PPAR? with IC50 of 3.3 nM in a cell-free assay, with at least 10- to 600-fold functional selectivity in cells with PPAR? versus PPAR? and PPAR?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3200