Learn More
Medchemexpress LLC HY-16578 10mg , GW9662 CAS:22978-25-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1657810MG
Encompass_Preferred
HY-16578 10mg GW9662 CAS: 22978-25-2 GW9662 is a potent and selective PPARγ antagonist with an IC50 of 3.3 nM, showing 10 and 1000-fold selectivity over PPARα and PPARδ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0976