Learn More
Aldevron gWiz™ Luc RG 10mg
Catalog No. 503073873
Encompass_Preferred
10mg dispense of gWiz Luciferase plasmid. The gWiz plasmids are designed for the highest level of gene expression in a wide range of mammalian cells and tissues. They contain a proprietary modified promoter, followed by intron A from the CMV immediate early gene, and a high-efficiency artificial transcription terminator. gWiz expression vectors are constructed using a plasmid backbone that has been extensively modified to achieve enhanced levels of transgene expression in mammalian cells.hese ready-to-use plasmids are endotoxin-free and can be applied directly to most in vitro and in vivo applications. Each gWiz plasmid is delivered in the buffer of your choice with delivery time of approximately 24 to 48 hours.
This product is subject to the following: Limited Use License
Catalog No. 50-307-3873 Supplier Aldevron Supplier No. 5001
10mg dispense of gWiz Luciferase plasmid. The gWiz plasmids are designed for the highest level of gene expression in a wide range of mammalian cells and tissues. They contain a proprietary modified promoter, followed by intron A from the CMV immediate early gene, and a high-efficiency artificial transcription terminator. gWiz expression vectors are constructed using a plasmid backbone that has been extensively modified to achieve enhanced levels of transgene expression in mammalian cells.hese ready-to-use plasmids are endotoxin-free and can be applied directly to most in vitro and in vivo applications. Each gWiz plasmid is delivered in the buffer of your choice with delivery time of approximately 24 to 48 hours.
This product is subject to the following: Limited Use License