Learn More
TARGETMOL CHEMICALS INC GX 201 5MG
SDP
Catalog No. 502578985
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GX 201 is a selective NaV1.7 inhibitor IC50 of < 3.2 nM for hNaV1.7. purity: 99%
Catalog No. 50-257-8985 Supplier TARGETMOL CHEMICALS INC Supplier No. T96475MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GX 201 is a selective NaV1.7 inhibitor IC50 of < 3.2 nM for hNaV1.7. purity: 99%