Learn More
TARGETMOL CHEMICALS INC GX-585 1MG
SDP
Catalog No. 502879220
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. GX-585 is a sulfonamide analog that is a Nav 1.7 channel inhibitor with analgesic activity for the study of neuropathic pain and inflammation. Purity 99.18%
Catalog No. 50-287-9220 Supplier TARGETMOL CHEMICALS INC Supplier No. T699151MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. GX-585 is a sulfonamide analog that is a Nav 1.7 channel inhibitor with analgesic activity for the study of neuropathic pain and inflammation. Purity 99.18%