Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gx-674 | 1432913-36-4 | MFCD32644319 | 97.3% | 534.95 | C21H13ClF2N6O3S2 | 50MG
SDP

Catalog No. 5000236617
Encompass_Preferred

GX-674 is a potent, state-dependent, isoform-selective antagonist of the voltage-gated sodium channel Nav1.7 developed for research applications. It demonstrates exceptionally high potency (IC50 0.1 nM at -40 mV) and is supplied as a solid with high reported purity suitable for pharmacology and electrophysiology studies.

  • Potent Nav1.7 antagonist with IC50 0.1 nM (-40 mV).
  • State-dependent, isoform-selective profile for targeted investigations.
  • High reported purity of 97.3% for consistent experimental performance.
  • Molecular weight 534.95; formula C21H13ClF2N6O3S2 for unambiguous identification.
  • Available in multiple package sizes, including 50 mg, for dosing flexibility.
  • Recommended storage: powder at -20°C; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-36617 Supplier Medchemexpress LLC Supplier No. HY12382550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GX-674 is a potent, state-dependent, isoform-selective antagonist of the voltage-gated sodium channel Nav1.7 developed for research applications. It demonstrates exceptionally high potency (IC50 0.1 nM at -40 mV) and is supplied as a solid with high reported purity suitable for pharmacology and electrophysiology studies.

  • Potent Nav1.7 antagonist with IC50 0.1 nM (-40 mV).
  • State-dependent, isoform-selective profile for targeted investigations.
  • High reported purity of 97.3% for consistent experimental performance.
  • Molecular weight 534.95; formula C21H13ClF2N6O3S2 for unambiguous identification.
  • Available in multiple package sizes, including 50 mg, for dosing flexibility.
  • Recommended storage: powder at -20°C; in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.