Learn More
Ion Torrent™ GX5™ Chip and Genexus™ Coupler

Description
The Ion Torrent GX5 Chip and Genexus Coupler includes two GX5 chips and two Genexus couplers, sufficient for two initializations of the Ion Torrent Genexus Integrated Sequencer. The GX5 Chip generates 12–15 million sequencing reads per lane, or 48–60 million reads per chip. The four-lane chip design provides maximum flexibility for sample processing, enabling sequencing of one to four lanes per run, with the in-use stability to utilize any remaining lanes within two weeks of the initial run.
As part of the Genexus Integrated Sequencer workflow, the GX5 Chip:
- Achieves 12–15 million reads per lane for 200–400 base-read libraries
- Maximizes flexibility with the ability to utilize one to four chip lanes per run
- Enables parallel processing of up to four compatible assays in a single run
- Supports intake variability of clinical research samples
The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.
Related Links:
Specifications
Specifications
| Content And Storage | • 2 GX5 chips • 2 Genexus couplers Store at room temperature. |
| For Use With (Equipment) | Genexus Integrated Sequencer |
| For Use With (Application) | Comparative Sequencing (Somatic Mutations - 10:50) |
| Product Line | GX5, Genexus |
| Product Type | Coupler |
| Quantity | 2 Runs |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.