Learn More
Medchemexpress LLC Gyki 52466 dihydrochloride | 2319722-40-0 | 99.8% | 366.24 | C17H17Cl2N3O2 | 50 MG
SDP
Catalog No. 5000247698
Encompass_Preferred
GYKI 52466 dihydrochloride is a selective, noncompetitive antagonist of AMPA and kainate ionotropic glutamate receptors used in preclinical neuroscience research. It is orally active, shows blood-brain barrier permeability, and has demonstrated anticonvulsant and neuroprotective effects in animal studies. Supplied as a solid for laboratory use.
  • Selective noncompetitive AMPA and kainate receptor antagonist.
  • Orally active with blood-brain barrier permeability.
  • Demonstrated anticonvulsant and neuroprotective effects in preclinical studies.
  • Supplied as a yellow to orange solid for laboratory use.
  • Store sealed, away from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-47698 Supplier Medchemexpress LLC Supplier No. HY103234A50MG
GYKI 52466 dihydrochloride is a selective, noncompetitive antagonist of AMPA and kainate ionotropic glutamate receptors used in preclinical neuroscience research. It is orally active, shows blood-brain barrier permeability, and has demonstrated anticonvulsant and neuroprotective effects in animal studies. Supplied as a solid for laboratory use.
  • Selective noncompetitive AMPA and kainate receptor antagonist.
  • Orally active with blood-brain barrier permeability.
  • Demonstrated anticonvulsant and neuroprotective effects in preclinical studies.
  • Supplied as a yellow to orange solid for laboratory use.
  • Store sealed, away from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month).