Learn More
Medchemexpress LLC Gyki 52466 dihydrochloride | 2319722-40-0 | 99.8% | 366.24 g·mol⁻1 | C17H17Cl2N3O2 | 100 MG
SDP
Catalog No. 5000247045
Encompass_Preferred
GYKI 52466 dihydrochloride is a research-grade small molecule antagonist selective for AMPA and kainate ionotropic glutamate receptors. It is orally active and noncompetitive, used in in vitro pharmacology, electrophysiology, and in vivo neuropharmacology studies.
  • Selective noncompetitive AMPA/kainate receptor antagonist.
  • Orally active with good blood-brain barrier permeability.
  • High chemical purity suitable for research applications.
  • Available in small milligram pack sizes for laboratory use.
  • Store sealed, away from moisture and light; follow recommended cold storage for solutions.
Catalog No. 50-002-47045 Supplier Medchemexpress LLC Supplier No. HY103234A100MG
GYKI 52466 dihydrochloride is a research-grade small molecule antagonist selective for AMPA and kainate ionotropic glutamate receptors. It is orally active and noncompetitive, used in in vitro pharmacology, electrophysiology, and in vivo neuropharmacology studies.
  • Selective noncompetitive AMPA/kainate receptor antagonist.
  • Orally active with good blood-brain barrier permeability.
  • High chemical purity suitable for research applications.
  • Available in small milligram pack sizes for laboratory use.
  • Store sealed, away from moisture and light; follow recommended cold storage for solutions.