Learn More
Medchemexpress LLC Gyki 52466 dihydrochloride | 2319722-40-0 | MFCD00153803 | 99.8% | 366.24 g/mol | C17H17Cl2N3O2 | 10 MG
SDP
Catalog No. 5000246220
Encompass_Preferred
GYKI 52466 dihydrochloride is a selective, noncompetitive antagonist of AMPA and kainate ionotropic glutamate receptors used in pharmacological research into anticonvulsant effects, neuroprotection, and excitatory neurotransmission studies.
  • Selective noncompetitive AMPA/kainate receptor antagonist.
  • Orally active with documented anticonvulsant activity.
  • High purity for research use (≈99.8%).
  • Supplied in small-molecule research quantities (e.g., 10 MG).
  • Molecular weight 366.24 g/mol; CAS 2319722-40-0.
  • Store sealed, away from moisture and light; recommended storage at 4°C (in solvent: -80°C/-20°C for long-term/short-term).
Catalog No. 50-002-46220 Supplier Medchemexpress LLC Supplier No. HY103234A10MG
GYKI 52466 dihydrochloride is a selective, noncompetitive antagonist of AMPA and kainate ionotropic glutamate receptors used in pharmacological research into anticonvulsant effects, neuroprotection, and excitatory neurotransmission studies.
  • Selective noncompetitive AMPA/kainate receptor antagonist.
  • Orally active with documented anticonvulsant activity.
  • High purity for research use (≈99.8%).
  • Supplied in small-molecule research quantities (e.g., 10 MG).
  • Molecular weight 366.24 g/mol; CAS 2319722-40-0.
  • Store sealed, away from moisture and light; recommended storage at 4°C (in solvent: -80°C/-20°C for long-term/short-term).