Learn More
Medchemexpress LLC Gyki 52466 dihydrochloride | 2319722-40-0 | MFCD00153803 | 99.8% | 366.24 g/mol | C17H17Cl2N3O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY103234A25MG
Encompass_Preferred
GYKI 52466 dihydrochloride is the dihydrochloride salt of a selective, non-competitive AMPA/kainate receptor antagonist used in neuroscience research for studies of excitatory neurotransmission, anticonvulsant activity, and neuroprotection. It is supplied as a high-purity, yellow to orange solid and is intended for research use only.
  • Selective non-competitive AMPA/kainate receptor antagonist.
  • Suitable for in vitro and in vivo neuroscience research.
  • High purity (~99.8%) compatible with analytical and biological assays.
  • Solid form, yellow to orange, with recommended sealed storage away from moisture and light.
  • Available in small research pack sizes for precise dosing and formulation.
Catalog No. 50-002-46219