Learn More
Cayman Chemical GYKI 52466 (hydrochloride), Purity: ≥98%, CAS: 192065-56-8, 10 mg
SDP
Catalog No. 502949956
Encompass_Preferred
An allosteric AMPA receptor antagonist; selectively inhibits AMPA-induced inward currents (IC50 = 7.5 µM) over NMDA- or GABA-induced inward currents in primary rat hippocampal neurons at 50 µM but also inhibits kainate-induced inward currents in the same cells (IC50 = 11 µM); reduces the amplitude of spontaneous EPSCs in the same cells at 10 µM; increases the latency to seizure onset and reduces mortality in a rat model of 4-AP-induced generalized tonic-clonic seizures at 25 and 50 mg/kg; prevents neuronal damage in the CA1 region of the hippocampus in a four-vessel occlusion-induced rat model of global ischemia-reperfusion injury at 30 mg/kg
Catalog No. 50-294-9956 Supplier Cayman Chemical Supplier No. 3441810 MG
An allosteric AMPA receptor antagonist; selectively inhibits AMPA-induced inward currents (IC50 = 7.5 µM) over NMDA- or GABA-induced inward currents in primary rat hippocampal neurons at 50 µM but also inhibits kainate-induced inward currents in the same cells (IC50 = 11 µM); reduces the amplitude of spontaneous EPSCs in the same cells at 10 µM; increases the latency to seizure onset and reduces mortality in a rat model of 4-AP-induced generalized tonic-clonic seizures at 25 and 50 mg/kg; prevents neuronal damage in the CA1 region of the hippocampus in a four-vessel occlusion-induced rat model of global ischemia-reperfusion injury at 30 mg/kg