Learn More
Medchemexpress LLC Gyki 53655 (hydrochloride) | 143692-48-2 | 98.4% | 388.85 g·mol⁻¹ | C19H21ClN4O3 | 100 MG
SDP
Catalog No. 5000247074
Encompass_Preferred
GYKI 53655 hydrochloride is a research-grade small molecule that acts as a non-competitive antagonist of AMPA and kainate receptors. It is supplied as a light yellow to yellow solid with reported purity of ~98.4% and molecular formula C19H21ClN4O3. Intended for in vitro research use only.
  • Non-competitive AMPA and kainate receptor antagonist.
  • High purity (~98.4%) for consistent experimental results.
  • Light yellow to yellow solid, soluble in DMSO and water with recommended handling precautions.
  • Recommended storage: solid at 4°C; prepared solutions at -80°C (up to 6 months) or -20°C (short term).
  • Available in multiple solid sizes, including 100 mg, for lab-scale studies.
Catalog No. 50-002-47074 Supplier Medchemexpress LLC Supplier No. HY103228100MG
GYKI 53655 hydrochloride is a research-grade small molecule that acts as a non-competitive antagonist of AMPA and kainate receptors. It is supplied as a light yellow to yellow solid with reported purity of ~98.4% and molecular formula C19H21ClN4O3. Intended for in vitro research use only.
  • Non-competitive AMPA and kainate receptor antagonist.
  • High purity (~98.4%) for consistent experimental results.
  • Light yellow to yellow solid, soluble in DMSO and water with recommended handling precautions.
  • Recommended storage: solid at 4°C; prepared solutions at -80°C (up to 6 months) or -20°C (short term).
  • Available in multiple solid sizes, including 100 mg, for lab-scale studies.