Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gyki 53655 (hydrochloride) | 143692-48-2 | MFCD01941362 | 98.4% | 388.85 g·mol⁻¹ | C19H21ClN4O3 | 1 ML
SDP

Catalog No. 5000247175
Encompass_Preferred

GYKI 53655 hydrochloride is a selective, non-competitive antagonist of AMPA receptors used in electrophysiology and receptor pharmacology research. The hydrochloride salt is available as a high-purity solid or as a ready-to-use 10 mM solution in DMSO.

  • Selective non-competitive AMPA and kainate receptor antagonist.
  • Available as 10 mM solution in DMSO (1 mL) or as solid doses.
  • High purity (~98.4%) suitable for in vitro studies.
  • Molecular weight 388.85 g·mol⁻¹ and formula C19H21ClN4O3.
  • Recommended storage: solid at 4°C; solution at -80°C for long-term storage.

Catalog No. 50-002-47175 Supplier Medchemexpress LLC Supplier No. HY10322810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GYKI 53655 hydrochloride is a selective, non-competitive antagonist of AMPA receptors used in electrophysiology and receptor pharmacology research. The hydrochloride salt is available as a high-purity solid or as a ready-to-use 10 mM solution in DMSO.

  • Selective non-competitive AMPA and kainate receptor antagonist.
  • Available as 10 mM solution in DMSO (1 mL) or as solid doses.
  • High purity (~98.4%) suitable for in vitro studies.
  • Molecular weight 388.85 g·mol⁻¹ and formula C19H21ClN4O3.
  • Recommended storage: solid at 4°C; solution at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.