Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GYKI 53655 hydrochloride | 143692-48-2 | MFCD01941362 | 98.4% | 388.85 g/mol | C19H21ClN4O3 | 50 MG
SDP

Catalog No. 5000246025
Encompass_Preferred

GYKI 53655 hydrochloride is the hydrochloride salt of GYKI 53655, a selective antagonist of alpha-amino-3-hydroxy-5-methylisoxazole-4-propionic acid (AMPA) receptors used as a research tool in neuropharmacology. The compound is supplied as a light yellow solid with high purity and documented solubility properties.

  • Selective antagonist of AMPA receptors for neuropharmacology research.
  • High purity suitable for in vitro pharmacology.
  • Soluble in DMSO and partially soluble in water with warming or sonication.
  • Supplied as a solid with recommended storage at 4°C, protected from moisture.
  • Molecular weight 388.85 g/mol and formula C19H21ClN4O3.

Catalog No. 50-002-46025 Supplier Medchemexpress LLC Supplier No. HY10322850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GYKI 53655 hydrochloride is the hydrochloride salt of GYKI 53655, a selective antagonist of alpha-amino-3-hydroxy-5-methylisoxazole-4-propionic acid (AMPA) receptors used as a research tool in neuropharmacology. The compound is supplied as a light yellow solid with high purity and documented solubility properties.

  • Selective antagonist of AMPA receptors for neuropharmacology research.
  • High purity suitable for in vitro pharmacology.
  • Soluble in DMSO and partially soluble in water with warming or sonication.
  • Supplied as a solid with recommended storage at 4°C, protected from moisture.
  • Molecular weight 388.85 g/mol and formula C19H21ClN4O3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.