Learn More
Medchemexpress LLC HY-103228 5mg Medchemexpress, GYKI53655 hydrochloride CAS:143692-48-2 Purity:>98%
SDP
Catalog No. 501968058
Encompass_Preferred
Medchemexpress, HY-103228 5mg GYKI53655 HCl CAS:143692-48-2 GYKI53655 hydrochloride is an α-amino-3-hydroxy-5-methylisoxazole-4-propionic acid (AMPA) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8058 Supplier Medchemexpress LLC Supplier No. HY1032285MG
Medchemexpress, HY-103228 5mg GYKI53655 HCl CAS:143692-48-2 GYKI53655 hydrochloride is an α-amino-3-hydroxy-5-methylisoxazole-4-propionic acid (AMPA) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.