Learn More
Cayman Chemical GymnmIc AcId I 1mg
SDP
Catalog No. 502939060
Encompass_Preferred
A triterpene glycoside; inhibits glucose-induced phosphorylation of serine 70 on S6K1 and serine 2448 on mTOR, caspase-3 activity, and apoptosis, as well as increases authophagy in MIN-6 pancreatic β cells at 5 UG/ml; has been used as a reference compound to compare the potency of plant-derived sweet taste inhibitors.
Catalog No. 50-293-9060 Supplier Cayman Chemical Supplier No. 284951 MG
A triterpene glycoside; inhibits glucose-induced phosphorylation of serine 70 on S6K1 and serine 2448 on mTOR, caspase-3 activity, and apoptosis, as well as increases authophagy in MIN-6 pancreatic β cells at 5 UG/ml; has been used as a reference compound to compare the potency of plant-derived sweet taste inhibitors.