Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2993MSM7P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: JC159. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Erythrocytes in any human tissues. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Formalin fixed membrane from a Hairy cell leukemia. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 39kDa.Chromosome: 4q31.22. Gene Symbol: GYPA. Gene ID: 2994. Ensemble: ENSG00000250361. Uniprot: NP_002091.
Catalog No. 50-188-4761