Learn More
Aviva Systems Biology Corp GYPA Antibody - N-terminal region (ARP63746_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63746_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GYPA. It was validated on Western Blot. . Alternate names: CD235a, GPA, GPErik, GPSAT, GpMiIII, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), MN, MNS, PAS-2ApplicationWB. Species reactivity: Human
Catalog No. 50-288-071