Learn More
Cayman Chemical Gypenoside IX, Purity: ≥95%, CAS: 80321-63-7, 25 mg
SDP
Catalog No. 502988932
Encompass_Preferred
A dammarane-type triterpene saponin; prevents increases in iNOS, IL-6, and TNF-α levels, as well as phosphorylation of NF-κB, IκB, p38 MAPK, and Akt, induced by LPS and TNF-α in rat C6 glial at 25 μM; reduces LPS-induced increases in NF-κB, IκB, p38 MAPK, and Akt phosphorylation in mouse brain cortex at 30 mg/kg per day
Catalog No. 50-298-8932 Supplier Cayman Chemical Supplier No. 2970525 MG
A dammarane-type triterpene saponin; prevents increases in iNOS, IL-6, and TNF-α levels, as well as phosphorylation of NF-κB, IκB, p38 MAPK, and Akt, induced by LPS and TNF-α in rat C6 glial at 25 μM; reduces LPS-induced increases in NF-κB, IκB, p38 MAPK, and Akt phosphorylation in mouse brain cortex at 30 mg/kg per day