Learn More
Medchemexpress LLC Gypenoside L | 94987-09-4 | 5 MG
SDP
Catalog No. 5000417305
Encompass_Preferred
Gypenoside L is a saponin found in Gynostemma pentaphyllum. It increases SA-β-galactosidase activity, promotes the production of senescence-associated secretory cytokines, and activates p38, ERK MAPK, and NF-κB pathways to induce senescence. It also exhibits anti-tumor and anti-inflammatory activities.
  • Saponin derived from Gynostemma pentaphyllum
  • Increases SA-β-galactosidase activity
  • Promotes production of senescence-associated secretory cytokines
  • Activates p38 and ERK MAPK pathways
  • Activates NF-κB pathway
  • Induces senescence
  • Exhibits anti-tumor activity
  • Exhibits anti-inflammatory activity
Catalog No. 50-004-17305 Supplier Medchemexpress LLC Supplier No. HYN82115MG
Gypenoside L is a saponin found in Gynostemma pentaphyllum. It increases SA-β-galactosidase activity, promotes the production of senescence-associated secretory cytokines, and activates p38, ERK MAPK, and NF-κB pathways to induce senescence. It also exhibits anti-tumor and anti-inflammatory activities.
  • Saponin derived from Gynostemma pentaphyllum
  • Increases SA-β-galactosidase activity
  • Promotes production of senescence-associated secretory cytokines
  • Activates p38 and ERK MAPK pathways
  • Activates NF-κB pathway
  • Induces senescence
  • Exhibits anti-tumor activity
  • Exhibits anti-inflammatory activity