Learn More
Medchemexpress LLC HY-N7678 1mg Medchemexpress, Gypenoside LXXV CAS:110261-98-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN76781MG
Encompass_Preferred
Medchemexpress, HY-N7678 1mg Gypenoside LXXV CAS:110261-98-8 Gypenoside LXXV, isolated from Gynostemma pentaphyllum, is one of the deglycosylated shapes of ginsenoside Rb1. Gypenoside LXXV significantly reduces cancer cell viability and displays an anti-cancer effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3007