Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gypenoside XIII | 80325-22-0 | 10 MG
SDP

Catalog No. 5000419327
Encompass_Preferred

Gypenoside XIII is a natural product belonging to the gypenosides, extracted from Gynostemma pentaphyllum. This compound exhibits various pharmacological properties and offers protective effects against cardiovascular diseases, particularly atherosclerosis. It has a molecular formula of C41H70O12, a molecular weight of 754.99, and a purity of 99.13%. Gypenoside XIII appears as a white to off-white solid. This product is for research use only.

  • Molecular formula: C41H70O12
  • Molecular weight: 754.99
  • High purity: 99.13%
  • Protective effects against cardiovascular diseases
  • For research use only

Catalog No. 50-004-19327 Supplier Medchemexpress LLC Supplier No. HYN688110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gypenoside XIII is a natural product belonging to the gypenosides, extracted from Gynostemma pentaphyllum. This compound exhibits various pharmacological properties and offers protective effects against cardiovascular diseases, particularly atherosclerosis. It has a molecular formula of C41H70O12, a molecular weight of 754.99, and a purity of 99.13%. Gypenoside XIII appears as a white to off-white solid. This product is for research use only.

  • Molecular formula: C41H70O12
  • Molecular weight: 754.99
  • High purity: 99.13%
  • Protective effects against cardiovascular diseases
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.