Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gypenoside XIII | 80325-22-0 | 5 MG
SDP

Catalog No. 5000418000
Encompass_Preferred

Gypenoside XIII is a natural product from the gypenosides family, extracted from Gynostemma pentaphyllum (Jiaogulan). It has a molecular weight of 754.99 and a formula of C41H70O12, appearing as a white to off-white solid with 99.13% purity. This triterpene is known for its various pharmacological properties.

  • Protects against cardiovascular diseases, particularly atherosclerosis.
  • Soluble in ethanol at 50 mg/mL, requiring sonication.
  • Storage at 4°C, protected from light.
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C, protected from light.

Catalog No. 50-004-18000 Supplier Medchemexpress LLC Supplier No. HYN68815MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gypenoside XIII is a natural product from the gypenosides family, extracted from Gynostemma pentaphyllum (Jiaogulan). It has a molecular weight of 754.99 and a formula of C41H70O12, appearing as a white to off-white solid with 99.13% purity. This triterpene is known for its various pharmacological properties.

  • Protects against cardiovascular diseases, particularly atherosclerosis.
  • Soluble in ethanol at 50 mg/mL, requiring sonication.
  • Storage at 4°C, protected from light.
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.