Learn More
Medchemexpress LLC Gypenoside XLIX | 94987-08-3 | 1047.23 | 20 MG
SDP
Catalog No. 5000415021
Encompass_Preferred
Gypenoside XLIX is a multifunctional bioactive compound isolated from Gynostemma pentaphyllum. It acts as a PPAR-α agonist and binds to SIRT1. It inhibits TLR4-mediated NF-κB signaling pathway activation, reduces ROS accumulation, and alleviates hepatic inflammatory injury. It targets SIRT1 to block YAP-NLRP3 activation, improving sepsis-induced cardiomyopathy. The compound is applicable to research related to acute liver injury, lung injury, cardiomyopathy, acute splenic injury, sepsis-associated encephalopathy, acute kidney injury, atherosclerosis, and chronic inflammation.
  • Acts as a PPAR-α agonist
  • Inhibits TLR4-mediated NF-κB signaling pathway
  • Reduces ROS accumulation
  • Alleviates hepatic inflammatory injury
  • Targets SIRT1 to block YAP-NLRP3 activation
  • Prevents acute kidney injury
Catalog No. 50-004-15021 Supplier Medchemexpress LLC Supplier No. HYN199020MG
Gypenoside XLIX is a multifunctional bioactive compound isolated from Gynostemma pentaphyllum. It acts as a PPAR-α agonist and binds to SIRT1. It inhibits TLR4-mediated NF-κB signaling pathway activation, reduces ROS accumulation, and alleviates hepatic inflammatory injury. It targets SIRT1 to block YAP-NLRP3 activation, improving sepsis-induced cardiomyopathy. The compound is applicable to research related to acute liver injury, lung injury, cardiomyopathy, acute splenic injury, sepsis-associated encephalopathy, acute kidney injury, atherosclerosis, and chronic inflammation.
  • Acts as a PPAR-α agonist
  • Inhibits TLR4-mediated NF-κB signaling pathway
  • Reduces ROS accumulation
  • Alleviates hepatic inflammatory injury
  • Targets SIRT1 to block YAP-NLRP3 activation
  • Prevents acute kidney injury