Learn More
Medchemexpress LLC Gypenoside XLVI | 94705-70-1 | 5 MG
SDP
Catalog No. 5000419207
Encompass_Preferred
Gypenoside XLVI is a major dammarane-type triterpenoid saponin extracted from *Gynostemma pentaphyllum*. It features a tetracyclic triterpene structure and demonstrates potent inhibitory activity against non-small cell lung carcinoma A549 cells.
  • Major dammarane-type triterpenoid saponin
  • Tetracyclic triterpene structure
  • Potent inhibitory activity against non-small cell lung carcinoma A549 cells
  • For research use only
Catalog No. 50-004-19207 Supplier Medchemexpress LLC Supplier No. HYN62525MG
Gypenoside XLVI is a major dammarane-type triterpenoid saponin extracted from *Gynostemma pentaphyllum*. It features a tetracyclic triterpene structure and demonstrates potent inhibitory activity against non-small cell lung carcinoma A549 cells.
  • Major dammarane-type triterpenoid saponin
  • Tetracyclic triterpene structure
  • Potent inhibitory activity against non-small cell lung carcinoma A549 cells
  • For research use only