Learn More
Cayman Chemical GypsogenIc AcId 2.5mg
SDP
Catalog No. 502912839
Encompass_Preferred
A triterpene acid with antibacterial and trypanocidal activities; active against the oral bacterial pathogens E. faecalis, S. salivarius, S. sanguinis, S. mitis, S. mutans, and S. sobrinus (MICs = 50-200 UG/ml); induces lysis of T. cruzi in isolated mouse blood (IC50 = 56.6 µM)
Catalog No. 50-291-2839 Supplier Cayman Chemical Supplier No. 360252.5 MG
A triterpene acid with antibacterial and trypanocidal activities; active against the oral bacterial pathogens E. faecalis, S. salivarius, S. sanguinis, S. mitis, S. mutans, and S. sobrinus (MICs = 50-200 UG/ml); induces lysis of T. cruzi in isolated mouse blood (IC50 = 56.6 µM)